Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Product Code. 11416598
Change view
Click to view available options
No. of Places:
384
Unit Size:
Each
Product Code. No. of Places unitSize
11416598 384 Each
1 options
Product Code. 11416598 Supplier Cole-Parmer™ Supplier No. undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these blocks for a variety of applications. Stuart™ Anodized Aluminum Blocks may be used in any combination.
  • For use with Stuart block heaters
  • Manufactured from anodized aluminum
  • Separate hole accommodates a thermometer if desired
Specifications
Description Holds 384 well plate
Holds 384 well plate
Material Anodized Aluminum
No. of Places 384
Type Block