missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Techne™ Aluminum Oxide for Fluidized Sand Baths
Click to view available options
For Use With (Equipment):
The Techne range of fluidized baths
Unit Size:
Each
Specifications
Specifications
| Color | Brown |
| For Use With (Equipment) | The Techne range of fluidized baths |
| Type | Alundum |
| Material | Fused Aluminum Oxide |
| Shape | Angular, Sharp Edges |
| Weight (Metric) | 25 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction