Learn More
Techne™ Dri-Block™ Aluminum Heating Block Insert
Product Code. 12163152
Click to view available options
No. of Places:
96
Unit Size:
Each
Product Code. 12163152
Brand: Techne™ 3662074
Only null left
Add to basket
Add to basket
Precision-engineered aluminum heating blocks offer excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
Specifications
Description No thermometer hole
Material Anodized Aluminum
No. of Places 96
Type Insert Block
Diameter (Metric) Well 1 mL