missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strychnine N-Oxide Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Strychnine N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Strychnine N-Oxide Hydrochloride |
| CAS | 139369-62-3 |
| Analyte or Component Names | Strychnine N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H22 N2 O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Strychnine N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C21H22N2O3.ClH/c24-18-10-16-19-13-9-17-21(6-7-23(17,25)11-12(13)5-8-26-16)14-3-1-2-4-15(14)22(18)20(19)21;/h1-5,13,16-17,19-20H,6-11H2;1H/t13-,16?,17-,19-,20?,21+,23?;/m0./s1 |
| SMILES | Cl.[O-][N+]12CC[C@@]34[C@@H]1C[C@@H]5[C@H]6C(CC(=O)N(C36)c7ccccc47)OCC=C5C2 |
| IUPAC Name | (4aR,5aS,8aS,15bR)-6-oxido-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-6-ium-14-one;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction