missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals
27.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Strontium titanium oxide is used as a diamond stimulant and also used as a strontium-90 in radioisotope thermoelectric generators. It is used in the electronics and electrical insulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2060°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water and most solvents. |
Formula Weight | 183.51 |
Percent Purity | ≥99% |
Odor | Odorless |
Density | 4.810 g/mL |
Assay | (metals basis) |
Chemical Name or Material | Strontium titanium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'