missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfide, 99.9% (metals basis)

Product Code. 11473850
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473850 50 g 50g
11483850 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473850 Supplier Thermo Scientific Alfa Aesar Supplier No. 089895.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-96-1 | SSr | 119.68 g/mol

Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-96-1
Molecular Formula SSr
Molecular Weight (g/mol) 119.68
MDL Number MFCD00049560
InChI Key ZEGFMFQPWDMMEP-UHFFFAOYSA-N
Synonym strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
PubChem CID 14820
IUPAC Name strontium(2+) sulfanediide
SMILES [S--].[Sr++]

Specifications

Melting Point >2000°C
Color Gray
Physical Form Powder
Quantity 50 g
Merck Index 14,8849
Solubility Information Slightly soluble in water; Decomposes in acids
Formula Weight 119.68
Percent Purity 99.9%
Odor Like rotten eggs
Sensitivity Hygroscopic
Density 3.70 g/mL
Assay (metals basis)
Chemical Name or Material Strontium sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.