missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Reagent Grade

Product Code. p-7050454
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11318827

Brand: Thermo Scientific Alfa Aesar 038373.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7759-02-6 | O4SSr | 183.68 g/mol

Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7759-02-6
ChEBI CHEBI:63041

Specifications

Melting Point 1600°C
Physical Form Crystalline
Quantity 25 g
Molecular Formula Cl2Mn
MDL Number MFCD00011114
Merck Index 14,8848
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
Solubility Information Soluble in dilute hydrochloric acid and nitric acid . Slightly soluble in water < 0.01g/L . Insoluble in ethanol and alkalis.
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) dichloride
Molecular Weight (g/mol) 125.84
PubChem CID 10313134
Formula Weight 183.68
Grade Reagent
Density 3.960 g/mL
Chemical Name or Material Strontium sulfate, For reagent grade
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.