missing translation for 'onlineSavingsMsg'
Learn More

Strontium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep

Product Code. 10132202 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Unit Size:
250mL
30mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132202 250 mL Poly Bottle 250mL
16331256 30 mL Triple-leached LDPE bottle 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10132202 Supplier SPEX Certiprep Supplier No. PLSR22T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Strontium Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 7440-24-6 , 7697-37-2 , 7732-18-5
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]

Specifications

Packaging Poly Bottle
Technique AA, ICP
Type Certified Reference Material
Quantity 250 mL
Grade Assurance
Chemical Name or Material Strontium Standard
compliance-icons
Product Identifier
  • 1000µg/ml strontium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P320-Specific treatment is urgent (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.