missing translation for 'onlineSavingsMsg'
Learn More

Strontium peroxide, 12.3% available oxygen

Product Code. 15462647
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462647 5 kg 5kg
15442647 250 g 250g
15452647 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462647 Supplier Thermo Scientific Alfa Aesar Supplier No. 046310.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-18-7 | O2Sr | 119.618 g/mol

Applications of Strontium Peroxide include pyrotechnics i.e. fireworks (red color), ammunitions (tracer/hunting), oxidizer, alarm rocket. It is also used in textiles for fibers bleaching and as an oxidizing agent for hair bleaching (professional).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-18-7
Molecular Formula O2Sr
Molecular Weight (g/mol) 119.618
MDL Number MFCD00084686
InChI Key UHCGLDSRFKGERO-UHFFFAOYSA-N
Synonym strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygen
PubChem CID 14807
IUPAC Name strontium;peroxide
SMILES [O-][O-].[Sr+2]

Specifications

Melting Point 215°C (decomposition)
Color Brown
Physical Form Powder
Quantity 5 kg
Linear Formula SrO2
UN Number UN1509
Merck Index 14,8847
Solubility Information Insoluble in water and slowly decomposes in water.
Formula Weight 119.62
Sensitivity Moisture sensitive
Chemical Name or Material Strontium peroxide, 12.3% available oxygen
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.