missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxide, 99.5% (metals basis), SrO ^=97%

Product Code. 11339219
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339219 100 g 100g
11349219 5 g 5g
11359219 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339219 Supplier Thermo Scientific Alfa Aesar Supplier No. 088220.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1314-11-0 | OSr | 103.619 g/mol

Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-11-0
Molecular Formula OSr
Molecular Weight (g/mol) 103.619
MDL Number MFCD00049558
InChI Key UFQXGXDIJMBKTC-UHFFFAOYSA-N
PubChem CID 518709
IUPAC Name oxostrontium
SMILES O=[Sr]

Specifications

Boiling Point ca 3000°C
Melting Point 2430°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in potassium hydroxide solution. Slightly soluble in alcohol. Insoluble in acetone and ether.
Formula Weight 103.62
Sensitivity Moisture sensitive
Density 4.7
Chemical Name or Material Strontium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.