missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxalate, 95%, Thermo Scientific Chemicals

Product Code. p-7050840
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11334477

Brand: Thermo Scientific Alfa Aesar 039573.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-95-9
Molecular Formula C2O4Sr
Molecular Weight (g/mol) 175.638
MDL Number MFCD00050803
InChI Key KQAGKTURZUKUCH-UHFFFAOYSA-L
Synonym strontium oxalate, unii-7iaw7e69ni, 7iaw7e69ni, strontium oxalate, src2o4, ethanedioic acid, strontium salt 1:1, oxalic acid, monostrontium salt, strontium 2+ ion oxalate, oxalic acid, strontium salt 1:1 8ci
PubChem CID 69947
IUPAC Name strontium;oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Sr+2]

Specifications

Quantity 500 g
UN Number UN3288
Solubility Information Soluble in hydrochloric acid and nitric acid
Formula Weight 175.64
Percent Purity 95%
Physical Form Powder or Lumps
Chemical Name or Material Strontium oxalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.