missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
29.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
250 g
1 kg
Description
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 570°C |
Physical Form | Crystalline |
Quantity | 250 g |
Assay Percent Range | 99.0% min. |
Solubility Information | Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid. |
IUPAC Name | strontium(2+) dinitrate |
Formula Weight | 211.63 |
Grade | ACS Reagent |
Sensitivity | Hygroscopic |
Density | 2.99 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'