missing translation for 'onlineSavingsMsg'
Learn More

Strontium molybdenum oxide, 99%

Product Code. 11383748
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383748 10 g 10g
11393748 50 g 50g
11303758 250 g 250g
11313758 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11383748 Supplier Thermo Scientific Alfa Aesar Supplier No. 040238.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13470-04-7 | SrMoO4

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13470-04-7
Molecular Formula SrMoO4
MDL Number MFCD00049550
Synonym Strontium molybdate

Specifications

Melting Point (decomposition)
Color Colorless
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (excluding Ba)
Solubility Information Insoluble in water. Soluble in acid
Formula Weight 247.56
Density 4.54
Chemical Name or Material Strontium molybdenum oxide, Ba < 0.3%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.