missing translation for 'onlineSavingsMsg'
Learn More

Strontium molybdenum oxide, 99%

Product Code. 11303758
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11303758

Brand: Thermo Scientific Alfa Aesar 040238.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13470-04-7 | SrMoO4

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13470-04-7
Molecular Formula SrMoO4
MDL Number MFCD00049550
Synonym Strontium molybdate

Specifications

Melting Point (decomposition)
Color Colorless
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (excluding Ba)
Solubility Information Insoluble in water. Soluble in acid
Formula Weight 247.56
Density 4.54
Chemical Name or Material Strontium molybdenum oxide, Ba < 0.3%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.