missing translation for 'onlineSavingsMsg'
Learn More

Strontium hydroxide octahydrate, tech.

Product Code. 11386598
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386598 2 kg 2kg
11306608 50 g 50g
11376598 500 g 500g
11396598 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11386598 Supplier Thermo Scientific Alfa Aesar Supplier No. 012199.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1311-10-0 | H18O10Sr | 265.75 g/mol

Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1311-10-0
Molecular Formula H18O10Sr
Molecular Weight (g/mol) 265.75
MDL Number MFCD00150395
InChI Key UJPWWRPNIRRCPJ-UHFFFAOYSA-L
IUPAC Name strontium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]

Specifications

Melting Point 100°C-8H2O
Physical Form Crystalline
Quantity 2 kg
Solubility Information Soluble in hot water and acid. Insoluble in acetone.
Formula Weight 265.76 (121.60 Anhydrous)
Grade Technical
Sensitivity Air sensitive; Hygroscopic
Density 1.9
Chemical Name or Material Strontium hydroxide octahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.