missing translation for 'onlineSavingsMsg'
Learn More

Strontium granules, 19mm (0.76in) & down, 99% (metals basis)

Product Code. 11378677
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378677 5 g 5g
11368677 25 g 25g
11388677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378677 Supplier Thermo Scientific Alfa Aesar Supplier No. 035789.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strontium granules, 99%, Sr, CAS Number-7440-24-6, atom, granules, 19mm 0.76in & down metals basis, estroncio, strontium, chunks, pieces, dendritic, pieces, yzs2rpe8le, strontium, elemental, strontium, distilled dendritic pieces, unii-yzs2rpe8le, 100g, (metals basis), CHEBI:33324, White to Yellow | CAS: 7440-24-6 | Sr | 87.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon with thin oil layer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-24-6
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
MDL Number MFCD00134060
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
Synonym strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]

Specifications

Color White to Yellow
Quantity 5 g
Particle Size 19mm (0.76 in.) & down
Sensitivity Air and moisture sensitive
UN Number UN3208
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Strontium granules
compliance-icons
Product Identifier
  • Strontium granules
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Strontium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.