missing translation for 'onlineSavingsMsg'
Learn More

Strontium fluoride, Optical Grade, Thermo Scientific™

Product Code. 11371628
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371628 2 g 2g
11381628 10 g 10g
11361628 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371628 Supplier Thermo Scientific Alfa Aesar Supplier No. 011020.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
PubChem CID 82210
SMILES [F-].[F-].[Sr++]

Specifications

Melting Point 1473°C
Physical Form Broken Crystals
Quantity 2 g
Merck Index 14,8842
IUPAC Name strontium(2+) difluoride
Formula Weight 125.62
Grade Optical
Odor Odorless
Refractive Index 1.442
Density 4.240 g/mL
Chemical Name or Material Strontium fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.