Learn More
Strontium fluoride, Puratronic™, 99.99% (metals basis)
Description
Strontium fluoride is used as an optical coating on lenses, and as a thermoluminescent dosimeter crystal. It also finds use as a water insoluble strontium source for use in oxygen-sensitive applications such as metal production. It serves both as a source of strontium and fluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 0.5 g |
| Assay Percent Range | 99.99%, metal basis |
| IUPAC Name | strontium;difluoride |
| Formula Weight | 125.62 |
| Chemical Name or Material | Strontium Fluoride |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.