missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride, ultra dry, 99.995% (metals basis)

Product Code. 11378567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378567 25 g 25g
11388567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378567 Supplier Thermo Scientific Alfa Aesar Supplier No. 035719.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10476-85-4 | SrCl2

Strontium chloride  is used as a source of redness in fireworks. Strontium chloride is often used as a red coloring agent in pyrotechnics. It is employed in small quantities in glass-making and metallurgy. The radioactive isotope strontium-89, used for the treatment of bone cancer, is usually administered in the form of strontium chloride. Sea water aquaria require small amounts of strontium chloride, which is consumed in the production of the exoskeletons of certain plankton. SrCl2 is useful in reducing tooth sensitivity by forming a barrier over microscopic tubules in the dentin containing nerve endings that have become exposed by gum recession. strontium chloride exposure induces parthenogenetic activation of oocytes[6] which is used in developmental biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
Molecular Formula SrCl2
MDL Number MFCD00011249

Specifications

Boiling Point 1250°C
Melting Point 873°C
Physical Form Beads
Quantity 25 g
Assay Percent Range 99.995% (metals basis)
Solubility Information It is soluble in water,slightly soluble in ethanol,acetone. Insoluble in ammonia.
Formula Weight 158.53
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052 g/mL
Chemical Name or Material Strontium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.