missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride, anhydrous, 95%

Product Code. p-7048958
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11356608

Brand: Thermo Scientific Alfa Aesar 012202.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10476-85-4

It is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
MDL Number MFCD00011249

Specifications

Boiling Point 1250°C
Melting Point 873°C
Color White
Physical Form Powder
Quantity 500 g
Merck Index 14,8840
Solubility Information Freely soluble in water. Very slightly soluble in ethanol and acetone.
Formula Weight 158.53
Percent Purity 95%
Odor Odorless
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052 g/mL
Chemical Name or Material Strontium chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.