missing translation for 'onlineSavingsMsg'
Learn More

Strontium Carbonate, Extra Pure, SLR, Fisher Chemical™

Product Code. 11438962 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438962 100 g 100g
11418862 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438962 Supplier Fisher Chemical Supplier No. S/8160/48
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Color White
Physical Form Solid
Quantity 100 g
Formula Weight 147.6182g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.