missing translation for 'onlineSavingsMsg'
Learn More

Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max

Product Code. 11387208
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387208 50 g 50g
11377208 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387208 Supplier Thermo Scientific Alfa Aesar Supplier No. 016791.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10476-81-0 | Br2Sr | 247.428 g/mol

Used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-81-0
Molecular Formula Br2Sr
Molecular Weight (g/mol) 247.428
MDL Number MFCD00049548
InChI Key YJPVTCSBVRMESK-UHFFFAOYSA-L
Synonym acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
PubChem CID 5360351
IUPAC Name strontium;dibromide
SMILES [Br-].[Br-].[Sr+2]

Specifications

Melting Point 643°C
Color White
Physical Form Granules
Quantity 50 g
Linear Formula SrBr2
Merck Index 14,8837
Solubility Information Soluble in water,carbinol,ethanol,amyl alcohol. Insoluble in ether.
Formula Weight 247.44
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.216 g/mL
Assay (metals basis excluding Ba), Ba 0.1% max.
Chemical Name or Material Strontium bromide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Strontium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Strontium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.