missing translation for 'onlineSavingsMsg'
Learn More

Strontium aluminate, 99.5% (metals basis)

Product Code. 11374597
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374597 25 g 25g
11364597 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374597 Supplier Thermo Scientific Alfa Aesar Supplier No. 039647.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12004-37-4 | SrAl2O4

Strontium aluminate acts as a photo luminescent phosphor with long persistence of phosphorescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-37-4
Molecular Formula SrAl2O4
MDL Number MFCD00054054
Synonym strontium aluminate, strontium;oxido oxo alumane, dioxo-2,4-dioxa-1,5-dialumina-3-strontapentane, strontium aluminate,-100 mesh trace metals basis

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 205.58
Density 3.559
Chemical Name or Material Strontium aluminate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.