missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)

Product Code. 11352088
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352088 25 g 25g
11362088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352088 Supplier Thermo Scientific Alfa Aesar Supplier No. 012990.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-94-2 | C4H6O4Sr | 205.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-94-2
Molecular Formula C4H6O4Sr
Molecular Weight (g/mol) 205.71
MDL Number MFCD00036392
InChI Key RXSHXLOMRZJCLB-UHFFFAOYSA-L
Synonym strontium acetate, strontium diacetate, acetic acid, strontium salt, strontium di acetate, unii-4sl32ymy7b, 4sl32ymy7b, acetic acid, strontium salt 2:1, acmc-1bn7n, strontium 2+ ion diacetate, ksc492a5d
PubChem CID 10987
SMILES [Sr++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 150°C (in -0.5H2O)
Physical Form Crystalline
Quantity 25 g
Linear Formula Sr(OOCCH3)2·xH2O
Beilstein 3692534
Merck Index 14,8836
IUPAC Name strontium(2+) diacetate
Formula Weight 205.71 (Anhydrous)
Percent Purity 99.9965%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Strontium acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.