missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hemihydrate, Reagent Grade

Product Code. 11376608
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376608 1 kg 1kg
11366608 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376608 Supplier Thermo Scientific Alfa Aesar Supplier No. 012203.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14692-29-6

Strontium acetate is used as a catalyst, as a chemical intermediate and in medicine. The primary application of strontium acetate is as an active component in toothpastes. This compound is used to desensitize teeth to extremes of hot and cold, sweets, acids, or touch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14692-29-6
MDL Number MFCD00198064

Specifications

Melting Point 150°C (in -0.5H2O)
Physical Form Crystalline
Quantity 1 kg
Merck Index 14,8836
Solubility Information Soluble in water and slightly soluble in alcohol.
Formula Weight 214.72 (205.71 Anhydrous)
Grade Reagent
Density 2.099
Chemical Name or Material Strontium acetate hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.