missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hemihydrate, 98%

Product Code. 11423207
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423207 100 g 100g
11433207 500 g 500g
11443207 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423207 Supplier Thermo Scientific Alfa Aesar Supplier No. A16149.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O

PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14692-29-6
Molecular Formula Sr(OOCCH3)2·0.5H2O
MDL Number MFCD00198064
Synonym Acetic acid strontium salt

Specifications

Melting Point 150°C (in -0.5H2O)
Quantity 100 g
Merck Index 14,8836
Solubility Information Soluble in water.
Formula Weight 214.72 (205.71 Anhydrous)
Percent Purity 98%
Chemical Name or Material Strontium acetate hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.