missing translation for 'onlineSavingsMsg'
Learn More

Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr)

Product Code. 11358648
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358648 25 g 25g
11368648 5 g 5g
11378648 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358648 Supplier Thermo Scientific Alfa Aesar Supplier No. 040719.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2457-02-5 | C6H8O4 | 144.13 g/mol

Strontium 2-ethylhexanoate is used in metal refining, plumbing, bleaching, engraving, plating, photography, disinfection, fertilizer manufacture, metal cleaning, and rust removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2457-02-5
MDL Number MFCD00070438

Specifications

Odor Mild
Molecular Formula C16H30O4Sr
Quantity 25 g
Synonym strontium 2-ethylhexanoate, strontium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, strontium salt 2:1, acmc-20ajmv, bis 2-ethylhexanoic acid strontium salt, hexanoic acid, 2-ethyl-, strontium salt, strontium 2+ bis 2-ethylhexanoate, hexanoic acid,2-ethyl-, strontium salt 2:1
Solubility Information Insoluble in water.
InChI Key DLWBHRIWCMOQKI-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sr+2]
IUPAC Name strontium;2-ethylhexanoate
Molecular Weight (g/mol) 374.03
PubChem CID 102822
Formula Weight 374.04
Concentration or Composition (by Analyte or Components) 40% in 2-ethylhexanoic acid (8 to 12% Sr)
Physical Form Liquid
Chemical Name or Material Strontium 2-ethylhexanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.