Learn More
STRN4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560404
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LCTFPTASEH GVPTSVAFTS TEPAHIVASF RSGDTVLYDM EVGSALLTLE SRGSSGPTQI NQVVSHPNQP LTITAHDDRG IRFLD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.
Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.Specifications
STRN4 | |
Polyclonal | |
Unconjugated | |
Strn4 | |
C80611; PPP2R6C; protein phosphatase 2 regulatory subunit B'''gamma; striatin 4; striatin, calmodulin binding protein 4; striatin-4; Strn4; Zin; zinedin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRL3 | |
Strn4 | |
Recombinant protein corresponding to Human STRN4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only