missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599830
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total STRN.
Striatin is found in all mammalian cells assayed and may be involved in vesicular transport, dendrite growth, and cellular signaling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also includes SG2NA and Zinedin. This antibody does not cross-react with SG2NA.
Specifications
| STRN | |
| Polyclonal | |
| Unconjugated | |
| STRN | |
| AU022939; D130055P19; PPP2R6A; protein phosphatase 2 regulatory subunit B'''alpha; SG2NA; Striatin; striatin, calmodulin binding protein; striatin, calmodulin-binding protein; STRN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 268980, 29149, 6801 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43815, O55106, P70483 | |
| STRN | |
| A synthesized peptide derived from human STRN(Accession O43815), corresponding to amino acid residues L109-R159. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction