missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553576
This item is not returnable.
View return policy
Description
Immunogen sequence: RALLGFSSDV TDREDDKNQD SVVNGTEAEV KETAMIAKSE LTDSASVLDN FKFLESAAAD FSDEDEDDDV DGREKSVIDT STIVRKKALP DSGEDRDTKE ALKEFDFLVT SEEGDNESRS AGDGTDWEKE DQC Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
Striatin is found in all mammalian cells assayed and may be involved in vesicular transport, dendrite growth, and cellular signaling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also includes SG2NA and Zinedin. This antibody does not cross-react with SG2NA.
Specifications
| STRN | |
| Polyclonal | |
| Unconjugated | |
| STRN | |
| AU022939; D130055P19; PPP2R6A; protein phosphatase 2 regulatory subunit B'''alpha; SG2NA; Striatin; striatin, calmodulin binding protein; striatin, calmodulin-binding protein; STRN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43815 | |
| STRN | |
| Recombinant protein corresponding to Human STRN. Recombinant protein control fragment (Product #RP-90664). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction