Learn More
Corning™ Stripettor™ Ultra Pipette Controller Accessory, Battery Set
Product Code. 17131855 Shop All Corning Products
Change view
Click to view available options
Quantity:
Set of 3
Unit Size:
Each
Product Code. Quantity Unit Size
17131855 Set of 3 Each
1 options
Product Code. 17131855 Supplier Corning™ Supplier No. 4994_BATTERY
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Battery set for the Stripettor Ultra Pipette Controller provide 8 hours of continuous use.
Battery set for Corning Stripettor Ultra Pipet Controller
TRUSTED_SUSTAINABILITY
Specifications
Battery Type NiHM
Type Battery Set
For Use With (Equipment) Stripettor Ultra Pipette Controller
Quantity Set of 3