Learn More
FAM40A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563837
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VLLNIMYLIV ETVHQECEGD KAEWRTMRQT FRAELGSPLY NNEPFAIMLF GMV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.Specifications
FAM40A | |
Polyclonal | |
Unconjugated | |
STRIP1 | |
6330569M22Rik; 6530401O14Rik; Fam40a; family with sequence similarity 40, member A; FAR11 factor arrest 11 homolog A; FAR11A; homolog of yeast FAR11 protein 1; hypothetical protein; Kiaa1761; Protein FAM40A; QnpA-18494; RP4-773N10.1; striatin interacting protein 1; striatin-interacting protein 1; striatin-interacting protein 1; LOW QUALITY PROTEIN: striatin-interacting protein 1; Strip1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VSL9 | |
STRIP1 | |
Recombinant protein corresponding to Human STRIP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only