missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRIP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93448-0.02ml
This item is not returnable.
View return policy
Description
STRIP1 Polyclonal antibody specifically detects STRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| STRIP1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| FAM40A, Family With Sequence Similarity 40, Member A, FAR11 Factor Arrest 11 Homolog A, FAR11 Factor Arrest 11 Homolog A (Yeast), FAR11A, Homolog Of Yeast FAR11 Protein 1, KIAA1761, Protein FAM40A, Striatin Interacting Protein 1, Striatin-Interacting Protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 743-837 of human STRIP1 (NP_149079.2). VRHRLNDDWAYGNDLDARPWDFQAEECALRANIERFNARRYDRAHSNPDFLPVDNCLQSVLGQRVDLPEDFQMNYDLWLEREVFSKPISWEELLQ | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85369 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction