missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin Sulfate, TRC

Product Code. 30005268
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005268 1 g 1g
18092430 10 g 10g
18022430 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005268 Supplier Toronto Research Chemicals Supplier No. TRCS6875001G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula 2 C21 H39 N7 O12 . 3 H2 O4 S
Molecular Weight (g/mol) 1457.38
Synonym Streptomycin sulfate, bis[N,N'-bis(aminoiminomethyl)-4-O-[5-deoxy-2-O-[2-deoxy-2-(methylamino)-a-l-glucopyranosyl]-3-C-formyl-a-l-lyxofuranosyl]-d-streptamine] trisulfate, D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-formyl-α-L-lyxofuranosyl-(1→4)-N1,N3-bis(aminoiminomethyl)-, sulfate (2:3), D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-formyl-α-L-lyxofuranosyl-(1→4)-N,N'-bis(aminoiminomethyl)-, sulfate (2:3) (salt) (9CI), Streptomycin, sulfate (2:3) (salt) (8CI), Ag Streptomycin, Agri-Strep, Agrimycin 17, Agrimycin sulfate, Ambistryn, Ambistryn S, FireWall (pesticide), Hortocyna, KeyStrepto, Plantomycin, Strepto, Streptobrettin, Streptomycin A sulfate, Streptomycin sesquisulfate, Streptomycin sesquisulphate, Streptomycin sulphate, Streptonex, Strycin, Vetstrep
IUPAC Name 1-[(1R,2R,3S,4R,5R,6S)-3-carbamimidamido-4-[(2R,3R,4R,5S)-3-[(2S,3S,4S,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;sulfuric acid
SMILES CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]3NC(=N)N)O[C@@H](C)[C@]2(O)C=O.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@H](O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]6NC(=N)N)O[C@@H](C)[C@]5(O)C=O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7+,8-,9-,10-,11+,12-,13-,14+,15+,16-,17-,18-,21+;;;/m00.../s1
Formula Weight 1456.4335
Percent Purity >95%
Chemical Name or Material Streptomycin sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.