missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin sulfate, Cell Culture Reagent

Product Code. 15423919
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423919 50 g 50g
15433919 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423919 Supplier Thermo Scientific Alfa Aesar Supplier No. J61299.18
  • ONLINE EXCLUSIVE PRICE
    £67.88
    £54.65 / 50g
    Save £13.23
    19% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity. | CAS: 3810-74-0 | C42H84N14O36S3 | 1457.376 g/mol

Inhibits initiation and causes misreading of rRNA in protein synthesis Streptomycin sulfate is used to study the mechanisms of streptomycin resistance. It is used to inhibit bacterial contamination in cell culture applications in association with penicillin. It is an antibiotic drug initially used for tuberculosis treatment. It inhibits protein synthesis by binding to 30S ribosomal subunit, which induces the misreading of the genetic code. It is involved in the initiation of translation of DNA, thereby results in death for a susceptible bacterium. Further, it controls bacterial and fungal diseases of certain fruit, vegetables, seed, ornamental crops and algae in ornamental ponds and aquaria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.
  • It inhibits the initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.

Application

  • Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance
  • This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture
  • Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis
  • This compound can be used as a protein synthesis inhibitor
  • It acts against gram-negative and gram-positive bacteria
  • Streptomycin sulfate also shows a fungicide activity
  • It can be used to remove nucleic acids from a biological extract
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
MDL Number MFCD00037023
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Type Streptomycin Sulfate
Physical Form Powder
Color White
Quantity 50 g
Beilstein 3894995
Merck Index 14,8826
Solubility Information Soluble in water.
Formula Weight 728.69
Grade Cell Culture
Chemical Name or Material Streptomycin sulfate
compliance-icons compliance-icons
Product Identifier
  • Streptomycin sulfate, Cell Culture Reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Streptomycin sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.