missing translation for 'onlineSavingsMsg'
Learn More

Melford Streptomycin Sulfate

Product Code. 30018816
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018816 50 g 50g
30018815 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018816 Supplier Melford Supplier No. S6200050.0
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streptomycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, disrupting the initiation complex and leading to misreading of mRNA.

Streptomycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, disrupting the initiation complex and leading to misreading of mRNA. This interference prevents proper translation and inhibits bacterial growth.

Streptomycin is employed in microbiology for selective bacterial isolation and antibiotic sensitivity testing to assess the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media aids in selectively isolating bacteria susceptible to this antibiotic.

Specifications

Chemical Name or Material Streptomycin Sulfate
CAS 3810-74-0
Quantity 50 g
Molecular Formula C42H78N14O24-3H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.