missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptomyces viridochromogenes pat Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117665
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Inactivates phosphinothricin (PPT) by transfer of an acetyl group from acetyl CoA. This enzyme is an effector of phosphinothricin tripeptide (PTT or bialaphos) resistance.
Specifications
| Streptomyces viridochromogenes pat | |
| Polyclonal | |
| Unconjugated | |
| pat; Phosphinothricin N-acetyltransferase; Phosphinothricin-resistance protein; PPT N-acetyltransferase | |
| Recombinant Streptomyces viridochromogenes Phosphinothricin N-acetyltransferase protein (1-183aa). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction