missing translation for 'onlineSavingsMsg'
Learn More

Streptococcus pneumoniae PS14 Antibody (5C7) - BSA Free, Novus Biologicals™

Product Code. 18383150 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383150 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383150 Supplier Novus Biologicals Supplier No. NBP274997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Streptococcus pneumoniae PS14 Monoclonal specifically detects Streptococcus pneumoniae PS14 in Bacteria samples. It is validated for N/A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus pneumoniae PS14
Classification Monoclonal
Clone 5C7
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias Capsular Polysaccharide Serotype 14 PS14, ST-14
Host Species Mouse
Immunogen Streptococcus pneumoniae PS14
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Streptococcus pneumoniae serotype 14 capsule
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.