missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (128/390)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183478
This item is not returnable.
View return policy
Description
Mouse anti Streptococcus pneumoniar antibody, clone 128/390 recognizes Streptococcus pneumoniae, a causative agent of respiratory disease in humans.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Specifications
| Streptococcus pneumoniae | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 128/390 | |
| Unconjugated | |
| Pneumococcus; s pneumoniae; S. pneumoniae | |
| Streptococcus pneumoniae whole cells. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction