missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus Group B Polyclonal Antibody, FITC

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173063
This item is not returnable.
View return policy
Description
The PA1-73063 antibody reacts with Streptococcus Group B. PA1-73063 has been successfully used in Immunofluorescence applications. The PA1-73063 antibody was raised against the type specific carbohydrate of group B Streptococcus.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
Specifications
| Streptococcus Group B | |
| Polyclonal | |
| FITC | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Type specific carbohydrate of group B Streptococcus. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction