Learn More
Invitrogen™ Streptococcus Group A Polyclonal Antibody, Biotin
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173062
Description
The PA1-73062 antibody reacts with Streptococcus Group A. This antibody does not cross-react with other Strep groups. PA1-73062 has been successfully used in immunofluorescence applications. The PA1-73062 antibody was raised against the type specific carbohydrate of group A Streptococcus.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
Specifications
| Streptococcus Group A | |
| Polyclonal | |
| Biotin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Type specific carbohydrate of group A Streptococcus. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.