missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus B Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB600-1398JF525
This item is not returnable.
View return policy
Description
Streptococcus B Polyclonal antibody specifically detects Streptococcus B in Bacteria samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Streptococcus B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Bacteria | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| Beta strep, Group b streptococci, Group b streptococcus, Strep b | |
| Streptococci, Group B | |
| 0.5 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction