missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Streptococcus B Polyclonal antibody specifically detects Streptococcus B in Bacteria samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | Streptococcus B |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Beta strep, Group b streptococci, Group b streptococcus, Strep b |
| Host Species | Rabbit |
| Immunogen | Streptococci, Group B |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?