missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptococcus Agalactiae Antibody (224/46), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62933AF594
This item is not returnable.
View return policy
Description
Streptococcus Agalactiae Monoclonal specifically detects Streptococcus Agalactiae in Bacteria samples. It is validated for ELISA.
Specifications
| Streptococcus Agalactiae | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Bacteria | |
| IgG3 |
| ELISA | |
| 224/46 | |
| ELISA | |
| s agalactiae, S. agalactiae | |
| Streptococcus agalactiae (NCTC 11360) | |
| 0.1 mL | |
| This product is specific for Streptococcus agalactiae a gram-positive streptococcus characterized by the presence of the group B Lancefield antigen. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction