missing translation for 'onlineSavingsMsg'
Learn More

Streptavidin, Streptomyces avidinii, Thermo Scientific Chemicals

Product Code. 15474809
Change view
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474809 10 mg 10mg
15464809 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474809 Supplier Thermo Scientific Alfa Aesar Supplier No. J62428.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptavidin, Streptomyces avidinii, CAS # 9013-20-1, is a crystalline protein isolated from the bacterium Streptomyces avidinii and shows biotin-binding activity.

Streptavidin is used as a tetrameric protein used to visualize biotin conjugated molecules in ELISA. Due to the strong affinity of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits. Streptavidin is widely used in molecular biology through its unique high affinity for the biotin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Streptavidin is a 60-kDa extracellular protein isolated from Streptomyces avidinii, and it is considered a tetrameric protein with a very high affinity to biotin
• It has four binding sites for biotin and has the ability to bind four molecules of biotin

Application

• This compound has a strong affinity for biotin and biotinylated molecules, and because of this, streptavidin is considered one of the most important components in laboratory research kits. Due to its affinity for biotin, it is widely used in several molecular biology techniques, including ELISA, DNA purification, enzyme immobilization, and flow cytometry
• Streptavidin is used to visualize biotin-conjugated molecules in enzyme-linked immunosorbent assay (ELISA)
• Streptavidin has been used in microbubble preparation, which is an important technique used in research
• It has been used extensively in bionanotechnology to prepare beads and nanoparticles
• It is used to immobilize proteins and oligonucleotides

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9013-20-1
Molecular Formula C14H17BrClNO2S
Molecular Weight (g/mol) 378.709
MDL Number MFCD00082035
InChI Key RTWACOLFHOBGCE-UHFFFAOYSA-N
PubChem CID 51062757
IUPAC Name 2-[(3-chlorophenyl)-piperidin-1-ium-1-ylidenemethyl]sulfanylacetic acid;bromide
SMILES C1CC[N+](=C(C2=CC(=CC=C2)Cl)SCC(=O)O)CC1.[Br-]

Specifications

Type Streptavidin
Physical Form Lyophilized Powder
Color White
Quantity 10 mg
Solubility Information Gives a clear solution at 5mg/ml in 0.1M NaCl. is soluble in water at 1mg/mL,or in phosphate buffered saline (PBS).
Formula Weight 55 kDa
Percent Purity 95%
Chemical Name or Material Streptavidin, Streptomyces avidinii

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.