missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (PE-Cy5.5)

Product Code. 10541295
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10541295

Brand: Invitrogen™ SA1018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin, (PE-Cy5.5)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate PE-Cy5.5
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 0.5 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, PE-Cy5.5 conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/694
Flow Cytometer Laser Lines 488 or 561, 488
Shipping Condition Wet Ice
Form Solution
Product Line Cy
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye PE-Cy5.5
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.