missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PE Conjugate

Product Code. 11500607
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11500607 250 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Azide
Product Code. 11500607 Supplier Invitrogen™ Supplier No. 12431787

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
Contains Azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser, Green Laser, Yellow-Green Laser

Emit

578 nm

Excite

488 - 561 nm

Reported Application

Flow Cytometric Analysis

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate PE (Phycoerythrin)
Quantity 250 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PE (Phycoerythrin)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.