missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin, Marina Blue™ conjugate
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Our streptavidin, Marina Blue™ conjugate is a bright, blue-fluorescent probe with excitation/emission ∼365/460 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.
Specifications
Specifications
| Conjugate | Marina Blue |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Shipping Condition | Room Temperature |
| Product Line | MARINA BLUE™ |
| Product Type | Conjugate |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction