missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin-Horseradish Peroxidase (HRP) Conjugate

Product Code. 10093212
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10093212 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10093212 Supplier Invitrogen™ Supplier No. SA10001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin-Horseradish Peroxidase (HRP) Conjugate

Streptavidin-HRP conjugate is typically used in the immunodetection of biotinylated proteins. This conjugate is suitable for use in immunoblotting, ELISA, immunomicroscopy. The amount of conjugate provided is sufficient for 25 western blots using 10 ml of working solution per reaction or 25 immunostaining reactions using 1 ml of working solution per reaction as appropriate.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.75 to 1.2 mg/mL
Conjugate HRP
Quantity 50 μL
Content And Storage Store in refrigerator (2—8°C).
Form Liquid
Product Line Invitrogen
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.