missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (FITC)

Product Code. 10757843
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10757843 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10757843 Supplier Invitrogen™ Supplier No. SA1001
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin, (FITC)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide
TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC (Fluorescein)
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 1 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, FITC conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/525
Flow Cytometer Laser Lines 488
Shipping Condition Wet Ice
Form Solution
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye FITC (Fluorescein)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.